accessibility menu, dialog, popup

UserName

CAS RN 615-41-8

ClI

CAS RN 615-41-8

IUPAC Name: 1-chloro-2-iodobenzene
Synonyms: 2-chloroiodobenzene
Molecular Weight (g/mol): 238.45
Molecular Formula: C6H4ClI
InChi Key: MPEOPBCQHNWNFB-UHFFFAOYSA-N
SMILES: ClC1=CC=CC=C1I
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)