accessibility menu, dialog, popup

UserName

CAS RN 61477-96-1

NaCH3NNONH(R)OHN(S)(S)SCH3CH3(R)NOOOOO

CAS RN 61477-96-1

IUPAC Name: sodium (2R,5S,6S)-6-[(2R)-2-[(4-ethyl-2,3-dioxopiperazine-1-carbonyl)amino]-2-phenylacetamido]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Synonyms: piperacillin
Molecular Weight (g/mol): 539.54
Molecular Formula: C23H26N5NaO7S
InChi Key: WCMIIGXFCMNQDS-RAMASXNTSA-M
SMILES: [Na+].CCN1CCN(C(=O)N[C@@H](C(=O)N[C@@H]2[C@@H]3SC(C)(C)[C@H](N3C2=O)C([O-])=O)C2=CC=CC=C2)C(=O)C1=O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Certifications/Compliance 
  • (1)
  • (1)
Color 
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)