accessibility menu, dialog, popup

UserName

ON

CAS RN 614-16-4

IUPAC Name: 3-oxo-3-phenylpropanenitrile
Synonyms: benzoylacetonitrile β-oxobenzenepropanenitrile
Molecular Weight (g/mol): 145.16
Molecular Formula: C9H7NO
InChi Key: ZJRCIQAMTAINCB-UHFFFAOYSA-N
SMILES: O=C(CC#N)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (3)
  • (3)
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)