accessibility menu, dialog, popup

UserName

CAS RN 614-10-8

CH3HNNHH2NS

CAS RN 614-10-8

IUPAC Name: [(2-methylphenyl)amino]thiourea
Synonyms: (2-methylphenyl)aminothiourea
Molecular Weight (g/mol): 181.26
Molecular Formula: C8H11N3S
InChi Key: YRABENKRWXQGMK-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1NNC(N)=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results