accessibility menu, dialog, popup

UserName

BrO

CAS RN 613-54-7

IUPAC Name: 2-bromo-1-(naphthalen-2-yl)ethan-1-one
Synonyms: 2-bromo-1-(naphthalen-2-yl)ethanone
Molecular Weight (g/mol): 249.11
Molecular Formula: C12H9BrO
InChi Key: YHXHHGDUANVQHE-UHFFFAOYSA-N
SMILES: BrCC(=O)C1=CC=C2C=CC=CC2=C1
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (1)
  • (4)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results