accessibility menu, dialog, popup

UserName

H3CNCH3

CAS RN 613-29-6

IUPAC Name: N,N-dibutylaniline
Synonyms: benzenamine, N,N-dibutyl-
Molecular Weight (g/mol): 205.35
Molecular Formula: C14H23N
InChi Key: FZPXKEPZZOEPGX-UHFFFAOYSA-N
SMILES: CCCCN(CCCC)C1=CC=CC=C1
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results