accessibility menu, dialog, popup

UserName

H3COHN

CAS RN 613-21-8

IUPAC Name: 2-methylquinolin-6-ol
Synonyms: 2-methylquinolin-6-ol
Molecular Weight (g/mol): 159.19
Molecular Formula: C10H9NO
InChi Key: USSQQASIZNTRAJ-UHFFFAOYSA-N
SMILES: CC1=CC=C2C=C(O)C=CC2=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results