accessibility menu, dialog, popup

UserName

H3CN

CAS RN 61204-03-3

IUPAC Name: 4-(4-heptylcyclohexyl)benzonitrile
Synonyms: 4-(4-heptylcyclohexyl)benzonitrile
Molecular Weight (g/mol): 283.46
Molecular Formula: C20H29N
InChi Key: NSGMZTNTQKRAFA-UHFFFAOYSA-N
SMILES: CCCCCCCC1CCC(CC1)C1=CC=C(C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results