accessibility menu, dialog, popup

UserName

CAS RN 6118-51-0

OOOO

CAS RN 6118-51-0

IUPAC Name: 4,10-dioxatricyclo[5.2.1.0²,⁶]dec-8-ene-3,5-dione
Synonyms: 4,10-dioxatricyclo[5.2.1.0²,⁶]dec-8-ene-3,5-dione
Molecular Weight (g/mol): 166.13
Molecular Formula: C8H6O4
InChi Key: QQYNRBAAQFZCLF-UHFFFAOYNA-N
SMILES: O=C1OC(=O)C2C3OC(C=C3)C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results