accessibility menu, dialog, popup

UserName

OH(R)OHO

CAS RN 611-71-2

IUPAC Name: (2R)-2-hydroxy-2-phenylacetic acid
Synonyms: (R)-mandelic acid mandelic acid, (S)-isomer (R)-mandelsaeure monosodium salt mandelic acid mandelsaeure (r)-isomer mandelic acid DL-amygdalic acid paramandelic acid monolithium salt mandelic acid mandelic acid, (S)-
Molecular Weight (g/mol): 152.15
Molecular Formula: C8H8O3
InChi Key: IWYDHOAUDWTVEP-SSDOTTSWSA-N
SMILES: O[C@@H](C(O)=O)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)
  • (3)
Percent Purity 
  • (3)
  • (2)
  • (2)
  • (1)
  • (3)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (3)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)