CAS RN 611-70-1
CAS RN 611-70-1
IUPAC Name:
2-methyl-1-phenylpropan-1-one
Synonyms:
2-methyl-1-phenylpropan-1-one
Molecular Weight (g/mol):
148.21
Molecular Formula:
C10H12O
InChi Key:
BSMGLVDZZMBWQB-UHFFFAOYSA-N
SMILES:
CC(C)C(=O)C1=CC=CC=C1
1
–
1
of
1
results
1
Isobutyrophenone, 97%
CAS: 611-70-1 Molecular Formula: C10H12O Molecular Weight (g/mol): 148.21 MDL Number: MFCD00008917 InChI Key: BSMGLVDZZMBWQB-UHFFFAOYSA-N Synonym: isobutyrophenone,isopropyl phenyl ketone,2-methylpropiophenone,1-propanone, 2-methyl-1-phenyl,phenyl isopropyl ketone,isopropyl phenyl keton,alpha-methylpropiophenone,unii-8l53972ntz,.alpha.-methylpropiophenone,2-methyl-1-phenyl-1-propanone PubChem CID: 69144 IUPAC Name: 2-methyl-1-phenylpropan-1-one SMILES: CC(C)C(=O)C1=CC=CC=C1