CAS RN 6100-74-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
1-(3-Hydroxy-4-methoxyphenyl)ethan-1-one (Acetoisovanillone), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Isoacetovanillone, TRC
CAS: 6100-74-9 Molecular Formula: C9 H10 O3 Molecular Weight (g/mol): 166.17 Synonym: 1-(3-Hydroxy-4-methoxyphenyl)ethanone,Acetophenone, 3'-hydroxy-4'-methoxy- (7CI,8CI),1-(3-Hydroxy-4-methoxyphenyl)ethanone,2-Methoxy-5-acetylphenol,3'-Hydroxy-4'-methoxyacetophenone,4'-Methoxy-3'-hydroxyacetophenone,5-Acetyl-2-methoxyphenol,Acetoisovanillone,Isoacetovanillone,NSC 30050,Diosmin Imp. A (EP) IUPAC Name: 1-(3-hydroxy-4-methoxyphenyl)ethanone SMILES: COc1ccc(cc1O)C(=O)C