accessibility menu, dialog, popup

UserName

H2OKKKOHOOOOOO

CAS RN 6100-05-6

IUPAC Name: tripotassium 2-hydroxypropane-1,2,3-tricarboxylate hydrate
Synonyms: tripotassium citrate hydrate
Molecular Weight (g/mol): 324.41
Molecular Formula: C6H7K3O8
InChi Key: PJAHUDTUZRZBKM-UHFFFAOYSA-K
SMILES: O.[K+].[K+].[K+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
Molecular Weight (g/mol) 
  • (8)
  • (2)
Physical Form 
  • (2)
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (2)
  • (2)
Grade 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (8)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)