accessibility menu, dialog, popup

UserName

CAS RN 61-54-1

H2NHN

CAS RN 61-54-1

IUPAC Name: 2-(1H-indol-3-yl)ethan-1-amine
Synonyms: tryptamine (amino-2 ethyl)-3 indole 2-indol-3-yl-aethylamin tryptamin
Molecular Weight (g/mol): 160.22
Molecular Formula: C10H12N2
InChi Key: APJYDQYYACXCRM-UHFFFAOYSA-N
SMILES: NCCC1=CNC2=CC=CC=C12
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)