accessibility menu, dialog, popup

UserName

CAS RN 60969-19-9

IUPAC Name: λ¹-thallanylium hexafluoro-λ⁵-phosphanuide
Synonyms: λ¹-thallanylium hexafluorophosphate
Molecular Weight (g/mol): 349.34
Molecular Formula: F6PTl
InChi Key: FRZBCOUMLHRKRT-UHFFFAOYSA-N
SMILES: [Tl+].F[P-](F)(F)(F)(F)F
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results