accessibility menu, dialog, popup

UserName

H3COOHN

CAS RN 608-08-2

IUPAC Name: 1H-indol-3-yl acetate
Synonyms: indole acetic acid indole-3-ol,acetate ester
Molecular Weight (g/mol): 175.19
Molecular Formula: C10H9NO2
InChi Key: JBOPQACSHPPKEP-UHFFFAOYSA-N
SMILES: CC(=O)OC1=CNC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (5)
  • (2)
  • (3)
Quantity 
  • (4)
  • (1)
  • (1)
  • (4)
Physical Form 
  • (3)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (5)
  • (2)
  • (3)
  • (4)
  • (1)
  • (1)
  • (4)
  • (3)
  • (2)