accessibility menu, dialog, popup

UserName

CAS RN 6066-82-6

IUPAC Name: 1-hydroxypyrrolidine-2,5-dione
Synonyms: N-hydroxysuccinimide
Molecular Weight (g/mol): 115.09
Molecular Formula: C4H5NO3
InChi Key: NQTADLQHYWFPDB-UHFFFAOYSA-N
SMILES: ON1C(=O)CCC1=O
Molecular Weight (g/mol) 
  • (3)
  • (6)
Percent Purity 
  • (3)
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (3)
  • (6)
  • (3)
  • (2)
  • (4)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (4)