accessibility menu, dialog, popup

UserName

CAS RN 606-68-8

N(R)(R)NN(R)(R)OOPPO(R)(R)(S)(S)NNOOOOOOOHHOONH2NH2OHHONaNa

CAS RN 606-68-8

IUPAC Name: disodium 1-[(2R,3R,4S,5R)-5-({[({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)(oxido)phosphoryl]oxy}methyl)-3,4-dihydroxyoxolan-2-yl]-3-carbamoyl-1λ⁵-pyridin-1-ylium
Synonyms: disodium nadh
Molecular Weight (g/mol): 708.4
Molecular Formula: C21H26N7Na2O14P2
InChi Key: GUIOTHWMFMEYPG-WUEGHLCSSA-M
SMILES: [Na+].[Na+].NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O
Molecular Weight (g/mol) 
  • (6)
  • (11)
Quantity 
  • (5)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (3)
Physical Form 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (2)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (9)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (3)
  • (6)
  • (11)
  • (5)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (3)