accessibility menu, dialog, popup

UserName

CAS RN 605-94-7

IUPAC Name: 2,3-dimethoxy-5-methylcyclohexa-2,5-diene-1,4-dione
Synonyms: coenzyme Q0 CoQ0 ubiquinone Q0 ubiquinone-O ubiquinone 0 ubiquinone-0
Molecular Weight (g/mol): 182.18
Molecular Formula: C9H10O4
InChi Key: UIXPTCZPFCVOQF-UHFFFAOYSA-N
SMILES: COC1=C(OC)C(=O)C(C)=CC1=O
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (5)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)