accessibility menu, dialog, popup

UserName

HClCH3HNNH2H3C

CAS RN 60481-51-8

IUPAC Name: hydrogen (3,4-dimethylphenyl)hydrazine chloride
Synonyms: hydrogen (3,4-dimethylphenyl)hydrazine chloride
Molecular Weight (g/mol): 172.66
Molecular Formula: C8H13ClN2
InChi Key: YYMIOVAEQIEPET-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CC1=CC=C(NN)C=C1C
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)