accessibility menu, dialog, popup

UserName

OO

CAS RN 604-59-1

IUPAC Name: 2-phenyl-4H-benzo[h]chromen-4-one
Synonyms: α-naphthoflavone
Molecular Weight (g/mol): 272.3
Molecular Formula: C19H12O2
InChi Key: VFMMPHCGEFXGIP-UHFFFAOYSA-N
SMILES: O=C1C=C(OC2=C1C=CC1=CC=CC=C21)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)