accessibility menu, dialog, popup

UserName

OHCl

CAS RN 604-44-4

IUPAC Name: 4-chloronaphthalen-1-ol
Synonyms: 4-chloro-1-naphthol
Molecular Weight (g/mol): 178.62
Molecular Formula: C10H7ClO
InChi Key: LVSPDZAGCBEQAV-UHFFFAOYSA-N
SMILES: OC1=C2C=CC=CC2=C(Cl)C=C1
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results