accessibility menu, dialog, popup

UserName

CAS RN 6038-19-3

IUPAC Name: hydrogen 3-aminothiolan-2-one chloride
Synonyms: hydrogen DL-homocysteine thiolactone chloride
Molecular Weight (g/mol): 153.62
Molecular Formula: C4H8ClNOS
InChi Key: ZSEGSUBKDDEALH-UHFFFAOYNA-N
SMILES: [H+].[Cl-].NC1CCSC1=O
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (1)
  • (4)
Physical Form 
  • (3)
  • (1)
Color 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (5)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)