accessibility menu, dialog, popup

UserName

H3COFOOHH2N

CAS RN 603105-80-2

IUPAC Name: 2-amino-3-(5-fluoro-2-methoxyphenyl)propanoic acid
Synonyms: 2-amino-3-(5-fluoro-2-methoxyphenyl)propanoic acid
Molecular Weight (g/mol): 213.21
Molecular Formula: C10H12FNO3
InChi Key: RVIQPUAYMLZLEM-UHFFFAOYNA-N
SMILES: COC1=C(CC(N)C(O)=O)C=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results