accessibility menu, dialog, popup

UserName

ClI

CAS RN 60274-60-4

IUPAC Name: 1-chloro-5-iodopentane
Synonyms: 1-chloro-5-iodopentane
Molecular Weight (g/mol): 232.49
Molecular Formula: C5H10ClI
InChi Key: GUUHKOCYWBEGGX-UHFFFAOYSA-N
SMILES: ClCCCCCI
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results