accessibility menu, dialog, popup

UserName

CAS RN 6020-87-7

IUPAC Name: 2-(N-methylcarbamimidamido)acetic acid hydrate
Synonyms: creatine hydrate
Molecular Weight (g/mol): 149.15
Molecular Formula: C4H11N3O3
InChi Key: MEJYXFHCRXAUIL-UHFFFAOYSA-N
SMILES: O.CN(CC(O)=O)C(N)=N
Percent Purity 
  • (3)
  • (6)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (2)
  • (3)
  • (6)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)