accessibility menu, dialog, popup

UserName

CAS RN 600-22-6

CH3OOCH3O

CAS RN 600-22-6

IUPAC Name: methyl 2-oxopropanoate
Synonyms: methyl-pyruvate
Molecular Weight (g/mol): 102.09
Molecular Formula: C4H6O3
InChi Key: CWKLZLBVOJRSOM-UHFFFAOYSA-N
SMILES: COC(=O)C(C)=O
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (6)