CAS RN 60-18-4
Thermo Scientific Chemicals L-Tyrosine, 99%
CAS: 60-18-4 Molecular Formula: C9H11NO3 Molecular Weight (g/mol): 181.19 MDL Number: MFCD00002606 InChI Key: OUYCCCASQSFEME-SVGMAFHSNA-N Synonym: l-tyrosine,tyrosine,s-tyrosine,p-tyrosine,h-tyr-oh,l-p-tyrosine,2s-2-amino-3-4-hydroxyphenyl propanoic acid,4-hydroxy-l-phenylalanine,l---tyrosine,tyrosine, l PubChem CID: 6057 ChEBI: CHEBI:17895 SMILES: N[C@@H](CC1=CC=C(O)C=C1)C(O)=O
Thermo Scientific Chemicals L-Tyrosine, 99+%
CAS: 60-18-4 Molecular Formula: C9H11NO3 Molecular Weight (g/mol): 181.19 MDL Number: MFCD00002606 InChI Key: OUYCCCASQSFEME-SVGMAFHSNA-N Synonym: l-tyrosine,tyrosine,s-tyrosine,p-tyrosine,h-tyr-oh,l-p-tyrosine,2s-2-amino-3-4-hydroxyphenyl propanoic acid,4-hydroxy-l-phenylalanine,l---tyrosine,tyrosine, l PubChem CID: 6057 ChEBI: CHEBI:17895 IUPAC Name: (2S)-2-amino-3-(4-hydroxyphenyl)propanoic acid SMILES: N[C@@H](CC1=CC=C(O)C=C1)C(O)=O
Thermo Scientific Chemicals L-Tyrosine, Cell Culture Reagent
CAS: 60-18-4 Molecular Formula: C9H11NO3 Molecular Weight (g/mol): 181.19 MDL Number: MFCD00002606 InChI Key: OUYCCCASQSFEME-SVGMAFHSNA-N Synonym: l-tyrosine,tyrosine,s-tyrosine,p-tyrosine,h-tyr-oh,l-p-tyrosine,2s-2-amino-3-4-hydroxyphenyl propanoic acid,4-hydroxy-l-phenylalanine,l---tyrosine,tyrosine, l PubChem CID: 6057 ChEBI: CHEBI:17895 SMILES: N[C@@H](CC1=CC=C(O)C=C1)C(O)=O
L-Tyrosine [Free Base], MP Biomedicals
CAS: 60-18-4 Molecular Formula: C9H11NO3 Molecular Weight (g/mol): 181.19 MDL Number: MFCD00002606 InChI Key: OUYCCCASQSFEME-SVGMAFHSNA-N Synonym: l-tyrosine,tyrosine,s-tyrosine,p-tyrosine,h-tyr-oh,l-p-tyrosine,2s-2-amino-3-4-hydroxyphenyl propanoic acid,4-hydroxy-l-phenylalanine,l---tyrosine,tyrosine, l PubChem CID: 6057 ChEBI: CHEBI:17895 IUPAC Name: (2S)-2-amino-3-(4-hydroxyphenyl)propanoic acid SMILES: N[C@@H](CC1=CC=C(O)C=C1)C(O)=O
L-Tyrosine, MP Biomedicals™
CAS: 60-18-4 Molecular Formula: C9H11NO3 Molecular Weight (g/mol): 181.19 MDL Number: MFCD00002606 InChI Key: OUYCCCASQSFEME-SVGMAFHSNA-N Synonym: l-tyrosine,tyrosine,s-tyrosine,p-tyrosine,h-tyr-oh,l-p-tyrosine,2s-2-amino-3-4-hydroxyphenyl propanoic acid,4-hydroxy-l-phenylalanine,l---tyrosine,tyrosine, l PubChem CID: 6057 ChEBI: CHEBI:17895 IUPAC Name: (2S)-2-amino-3-(4-hydroxyphenyl)propanoic acid SMILES: N[C@@H](CC1=CC=C(O)C=C1)C(O)=O
(S)-(-)-Tyrosine, Sigma-Aldrich™
CAS: 60-18-4 Grade: Research Molecular Weight (g/mol): 181.19
l-Tyrosine, Millipore™
CAS: 60-18-4
Tyrosine, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
L-Tyrosine, TRC
CAS: 60-18-4 Molecular Formula: C9 H11 N O3 Molecular Weight (g/mol): 181.19 Synonym: L-Tyrosine,Tyrosine, L- (8CI),(-)-α-Amino-p-hydroxyhydrocinnamic acid,(2S)-2-Amino-3-(4-hydroxyphenyl)propanoic acid,(S)-2-Amino-3-(4-hydroxyphenyl)propanoic acid,(S)-Tyrosine,(S)-α-Amino-4-hydroxybenzenepropanoic acid,4-(2-Amino-2-carboxyethyl)phenol,Benzenepropanoic acid, α-amino-4-hydroxy-, (S)-,H-L-Tyr-OH,L(-)-β-(p-Hydroxyphenyl)alanine,L-(-)-Tyrosine,L-4-Hydroxyphenylalanine,L-Phenylalanine, 4-hydroxy-,L-Tyr,L-p-Tyrosine,NSC 82624,NSC 9973,Propanoic acid, 2-amino-3-(4-hydroxyphenyl)-, (S)-,Therigon,Tyr,Tyrosine,p-Hydroxyphenylalanine,p-Tyrosine,β-(p-Hydroxyphenyl)alanine,Levodopa Imp. B (EP) IUPAC Name: (2S)-2-amino-3-(4-hydroxyphenyl)propanoic acid SMILES: N[C@@H](Cc1ccc(O)cc1)C(=O)O
L-Tyrosine, MedChemExpress
MedChemExpress L-Tyrosine is a non-essential amino acid which can inhibit citrate synthase activity in the posterior cortex.
Tyrosine (L-Tyrosine), LoGiCal
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.