accessibility menu, dialog, popup

UserName

H3CNCH3NN(E/Z)

CAS RN 60-11-7

IUPAC Name: N,N-dimethyl-4-(2-phenyldiazen-1-yl)aniline
Synonyms: fat yellow DMAB iketon yellow extra petrol yellow WT waxoline yellow ads fat yellow AD OO fat yellow ES extra oil yellow 20 oil yellow 2G oil yellow DN
Molecular Weight (g/mol): 225.3
Molecular Formula: C14H15N3
InChi Key: JCYPECIVGRXBMO-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (8)
Quantity 
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
Grade 
  • (2)
  • (3)
Color 
  • (2)
pH 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (8)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)