accessibility menu, dialog, popup

UserName

SNHHNNN(E/Z)

CAS RN 60-10-6

IUPAC Name: 3-(phenylamino)-1-(phenylimino)thiourea
Synonyms: dithizone
Molecular Weight (g/mol): 256.33
Molecular Formula: C13H12N4S
InChi Key: UOFGSWVZMUXXIY-UHFFFAOYSA-N
SMILES: S=C(NNC1=CC=CC=C1)N=NC1=CC=CC=C1
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (4)
Grade 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (4)
  • (3)
  • (3)