accessibility menu, dialog, popup

UserName

CAS RN 5993-91-9

H2NHNNHClHCl

CAS RN 5993-91-9

IUPAC Name: dihydrogen 1-(1H-1,3-benzodiazol-2-yl)methanamine hydrate dichloride
Synonyms: dihydrogen 1-(1H-1,3-benzodiazol-2-yl)methanamine hydrate dichloride
Molecular Weight (g/mol): 238.11
Molecular Formula: C8H13Cl2N3O
InChi Key: QHEZHWNFSVYUGT-UHFFFAOYSA-N
SMILES: [H+].[H+].O.[Cl-].[Cl-].NCC1=NC2=CC=CC=C2N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results