CAS RN 598-18-5
CAS RN 598-18-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Bromo-1-propanol, TRC
CAS: 598-18-5 Molecular Formula: C3H7BrO Molecular Weight (g/mol): 138.99 Synonym: (±)-2-Bromo-1-propanol,1-Hydroxy-2-bromopropane,2-Bromo-1-propanol,2-Bromopropanol,2-Hydroxy-1-methylethyl bromide IUPAC Name: 2-bromopropan-1-ol SMILES: OCC(Br)C