accessibility menu, dialog, popup

UserName

CAS RN 59748-90-2

HOOBrCl

CAS RN 59748-90-2

IUPAC Name: 4-bromo-2-chlorobenzoic acid
Synonyms: 4-bromo-2-chlorobenzoic acid
Molecular Weight (g/mol): 235.46
Molecular Formula: C7H4BrClO2
InChi Key: JAVZWSOFJKYSDY-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(Br)C=C1Cl
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (3)
Quantity 
  • (3)
  • (1)
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)