Learn More
4-Bromo-2-chlorobenzoic Acid, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | 4-Bromo-2-chlorobenzoic Acid |
| CAS | 59748-90-2 |
| Molecular Formula | C7H4BrClO2 |
| Quantity | 1 g |
| Synonym | 2-Chloro-4-bromobenzoic Acid, 4-Bromo-2-chloro-benzoic Acid |
| InChI Formula | InChI=1S/C7H4BrClO2/c8-4-1-2-5(7(10)11)6(9)3-4/h1-3H,(H,10,11) |
| SMILES | OC(=O)c1ccc(Br)cc1Cl |
| IUPAC Name | 4-bromo-2-chlorobenzoic acid |
| Molecular Weight (g/mol) | 235.46 |
| Formula Weight | 233.9083 |