accessibility menu, dialog, popup

UserName

H2NOHO

CAS RN 5959-52-4

IUPAC Name: 3-aminonaphthalene-2-carboxylic acid
Synonyms: 3-amino-2-naphthoic acid
Molecular Weight (g/mol): 187.2
Molecular Formula: C11H9NO2
InChi Key: XFXOLBNQYFRSLQ-UHFFFAOYSA-N
SMILES: NC1=C(C=C2C=CC=CC2=C1)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)