accessibility menu, dialog, popup

UserName

CAS RN 59575-91-6

NH2NHCl

CAS RN 59575-91-6

IUPAC Name: 1-phenyl-1-(pyridin-2-yl)methanamine hydrochloride
Synonyms: 1-phenyl-1-(pyridin-2-yl)methanamine hydrochloride
Molecular Weight (g/mol): 220.7
Molecular Formula: C12H13ClN2
InChi Key: GVGSFONXPJCBIS-UHFFFAOYNA-N
SMILES: Cl.NC(C1=CC=CC=C1)C1=CC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results