accessibility menu, dialog, popup

UserName

H3CH3CCH3OHO

CAS RN 595-37-9

IUPAC Name: 2,2-dimethylbutanoic acid
Synonyms: 2,2-dimethyl butyric acid dimethyl ethyl acetic acid DMB
Molecular Weight (g/mol): 116.16
Molecular Formula: C6H12O2
InChi Key: VUAXHMVRKOTJKP-UHFFFAOYSA-N
SMILES: CCC(C)(C)C(O)=O
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results