accessibility menu, dialog, popup

UserName

CAS RN 59433-50-0

CH3H3CCH3OONOHO

CAS RN 59433-50-0

IUPAC Name: 1-[(tert-butoxy)carbonyl]pyrrolidine-2-carboxylic acid
Synonyms: 1-(tert-butoxycarbonyl)pyrrolidine-2-carboxylic acid
Molecular Weight (g/mol): 215.25
Molecular Formula: C10H17NO4
InChi Key: ZQEBQGAAWMOMAI-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CCCC1C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results