accessibility menu, dialog, popup

UserName

CAS RN 594-70-7

CH3H3CCH3NOO

CAS RN 594-70-7

IUPAC Name: 2-methyl-2-nitropropane
Synonyms: 2-methyl-2-nitropropane
Molecular Weight (g/mol): 103.12
Molecular Formula: C4H9NO2
InChi Key: AIMREYQYBFBEGQ-UHFFFAOYSA-N
SMILES: CC(C)(C)[N+]([O-])=O
Boiling Point 
  • (4)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (4)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)