CAS RN 594-48-9
CAS RN 594-48-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2,2-Dibromopropanoic Acid, TRC
CAS: 594-48-9 Molecular Formula: C3H4Br2O2 Molecular Weight (g/mol): 231.87 Synonym: 2,2-Dibromopropionic Acid; IUPAC Name: 2,2-dibromopropanoic acid SMILES: CC(Br)(Br)C(=O)O