accessibility menu, dialog, popup

UserName

CH3CH3LiH3C

CAS RN 594-19-4

IUPAC Name: lithium(1+) 2-methylpropan-2-ide
Synonyms: tert-butyllithium
Molecular Weight (g/mol): 64.06
Molecular Formula: C4H9Li
InChi Key: UBJFKNSINUCEAL-UHFFFAOYSA-N
SMILES: [Li+].C[C-](C)C
Quantity 
  • (2)
  • (1)
Boiling Point 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results