accessibility menu, dialog, popup

UserName

CH3OOSSOOCl

CAS RN 59337-92-7

IUPAC Name: methyl 3-(chlorosulfonyl)thiophene-2-carboxylate
Synonyms: methyl 3-(chlorosulfonyl)thiophene-2-carboxylate
Molecular Weight (g/mol): 240.67
Molecular Formula: C6H5ClO4S2
InChi Key: PJVJBDAUWILEOG-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(C=CS1)S(Cl)(=O)=O
Molecular Weight (g/mol) 
  • (6)
  • (4)
Quantity 
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (3)
  • (6)
  • (4)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)