accessibility menu, dialog, popup

UserName

CAS RN 59277-89-3

NNNHNOH2NOOH

CAS RN 59277-89-3

IUPAC Name: 2-amino-9-[(2-hydroxyethoxy)methyl]-6,9-dihydro-3H-purin-6-one
Synonyms: aciclovir aciclostad cicloferon aciclovir grin brand ophthalmic form aciclovir warner lambert brand of aciclovir aciclovir azupharma brand virax-puren yamanouchi brand of aciclovir rentschler brand of aciclovir sodium salt
Molecular Weight (g/mol): 225.21
Molecular Formula: C8H11N5O3
InChi Key: MKUXAQIIEYXACX-UHFFFAOYSA-N
SMILES: NC1=NC(=O)C2=C(N1)N(COCCO)C=N2
Molecular Weight (g/mol) 
  • (9)
  • (6)
Percent Purity 
  • (3)
  • (1)
  • (5)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
Physical Form 
  • (3)
  • (3)
Certifications/Compliance 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (3)
  • (3)
  • (9)
  • (6)
  • (3)
  • (1)
  • (5)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (3)
  • (3)
  • (2)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)