accessibility menu, dialog, popup

UserName

CAS RN 59247-52-8

H3CCH3OOBr

CAS RN 59247-52-8

IUPAC Name: propan-2-yl 2-bromobenzoate
Synonyms: isopropyl 2-bromobenzoate
Molecular Weight (g/mol): 243.1
Molecular Formula: C10H11BrO2
InChi Key: QTCRCHHNKWSPHB-UHFFFAOYSA-N
SMILES: CC(C)OC(=O)C1=CC=CC=C1Br
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results