CAS RN 5922-37-2
CAS RN 5922-37-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Desmethyl Itopride Hydrochloride, TRC
CAS: 5922-37-2 Molecular Formula: C19 H24 N2 O4 . Cl H Molecular Weight (g/mol): 380.86 Synonym: 3,4-dimethoxy-N-(4-(2-(methylamino)ethoxy)benzyl)benzamide, hydrochloride (1:1) IUPAC Name: 3,4-dimethoxy-N-[[4-[2-(methylamino)ethoxy]phenyl]methyl]benzamide;hydrochloride SMILES: Cl.CNCCOc1ccc(CNC(=O)c2ccc(OC)c(OC)c2)cc1