accessibility menu, dialog, popup

UserName

H3CCH3CH3

CAS RN 592-27-8

IUPAC Name: 2-methylheptane
Synonyms: 2-methylheptane
Molecular Weight (g/mol): 114.23
Molecular Formula: C8H18
InChi Key: JVSWJIKNEAIKJW-UHFFFAOYSA-N
SMILES: CCCCCC(C)C
Molecular Weight (g/mol) 
  • (3)
  • (4)
Boiling Point 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results