CAS RN 59107-99-2
IUPAC Name:
1,3-dimethyl 2-(2-methylbutyl)propanedioate
Synonyms:
1,3-dimethyl 2-(2-methylbutyl)propanedioate
Molecular Weight (g/mol):
202.25
Molecular Formula:
C10H18O4
InChi Key:
NYHSCTJAXMPYJG-UHFFFAOYNA-N
SMILES:
CCC(C)CC(C(=O)OC)C(=O)OC
1
–
1
of
1
results
1
Dimethyl (2-methylbutyl)malonate, 97%, Thermo Scientific™
CAS: 59107-99-2 Molecular Formula: C10H18O4 Molecular Weight (g/mol): 202.25 MDL Number: MFCD08436117 InChI Key: NYHSCTJAXMPYJG-UHFFFAOYSA-N Synonym: dimethyl 2-methylbutyl malonate,dimethyl 2-2-methylbutyl malonate,dimethyl 2-methylbutyl propanedioate,dimethyl 2-2-methylbutyl propanedioate,2-methylbutyl malonic acid dimethyl ester,1,3-dimethyl 2-2-methylbutyl propanedioate PubChem CID: 46779063 IUPAC Name: dimethyl 2-(2-methylbutyl)propanedioate SMILES: CCC(C)CC(C(=O)OC)C(=O)OC