accessibility menu, dialog, popup

UserName

CAS RN 591-31-1

CH3OO

CAS RN 591-31-1

IUPAC Name: 3-methoxybenzaldehyde
Synonyms: benzaldehyde, 3-methoxy-
Molecular Weight (g/mol): 136.15
Molecular Formula: C8H8O2
InChi Key: WMPDAIZRQDCGFH-UHFFFAOYSA-N
SMILES: COC1=CC=CC(C=O)=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)