CAS RN 591-07-1
CAS RN 591-07-1
IUPAC Name:
acetylurea
Synonyms:
acetamide, N-(aminocarbonyl)-
Molecular Weight (g/mol):
102.09
Molecular Formula:
C3H6N2O2
InChi Key:
GKRZNOGGALENQJ-UHFFFAOYSA-N
SMILES:
CC(=O)NC(N)=O
1
–
2
of
2
results
1
Acetylurea, TRC
CAS: 591-07-1 Molecular Formula: C3H6N2O2 Molecular Weight (g/mol): 102.09 Synonym: N-(Aminocarbonyl)acetamide,1-Acetylurea,Acetylcarbamide,Monoacetylurea,N-Acetylurea,NSC 2766 IUPAC Name: N-carbamoylacetamide SMILES: CC(=O)NC(=O)N
2
Acetylurea, 98%, Thermo Scientific™
CAS: 591-07-1 Molecular Formula: C3H6N2O2 Molecular Weight (g/mol): 102.093 MDL Number: MFCD00038140 InChI Key: GKRZNOGGALENQJ-UHFFFAOYSA-N Synonym: acetylurea,acetamide, n-aminocarbonyl,n-acetylurea,acetylcarbamide,monoacetylurea,1-acetylurea,acetic acid, ureide,n-aminocarbonyl acetamide,urea, acetyl,unii-8kru31i919 PubChem CID: 68956 IUPAC Name: N-carbamoylacetamide SMILES: CC(=O)NC(=O)N