CAS RN 590371-81-6
IUPAC Name:
2-chloro-4-(trifluoromethyl)pyridine-3-carboxylic acid
Synonyms:
2-chloro-4-(trifluoromethyl)pyridine-3-carboxylic acid
Molecular Weight (g/mol):
225.55
Molecular Formula:
C7H3ClF3NO2
InChi Key:
FZIJHEOXLKCIRZ-UHFFFAOYSA-N
SMILES:
OC(=O)C1=C(C=CN=C1Cl)C(F)(F)F
1
–
2
of
2
results
1
2-Chloro-4-(trifluoromethyl)nicotinic acid, 97%, Thermo Scientific™
CAS: 590371-81-6 Molecular Formula: C7H3ClF3NO2 Molecular Weight (g/mol): 225.55 MDL Number: MFCD08458125 InChI Key: FZIJHEOXLKCIRZ-UHFFFAOYSA-N Synonym: 2-chloro-4-trifluoromethyl nicotinic acid,2-chloro-4-trifluoromethyl pyridine-3-carboxylic acid,3-carboxy-2-chloro-4-trifluoromethyl pyridine,acmc-1az7h,ksc269c2b,3-pyridinecarboxylicacid, 2-chloro-4-trifluoromethyl,3-pyridinecarboxylic acid, 2-chloro-4-trifluoromethyl,2-chloro-4-trifluoromethyl-3-pyridinecarboxylic acid PubChem CID: 11770138 IUPAC Name: 2-chloro-4-(trifluoromethyl)pyridine-3-carboxylic acid SMILES: OC(=O)C1=C(C=CN=C1Cl)C(F)(F)F